Order Entry
Belgium
ContactUsLinkComponent
Barium chloride 1 g/l, VOLUSOL® for hydrometry
Barium chloride 1 g/l, VOLUSOL® for hydrometry
Catalog # 30594.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Barium chloride 1 g/l, VOLUSOL® for hydrometry
Catalog # 30594.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (complexometric, calc. on the ignition residue):
    Min 99 %
  • Loss on ignition (1000 °C):
    Max 1.0 %

Specification Test Results

About this item

 :
 :208.24 g/mol
Storage Temperature:Ambient
EINECS:241-234-5
UN:3287
ADR:6.1,III
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445