Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Perfluorodecanoic acid 50 µg/ml in methanol
Organic reference standard, Perfluorodecanoic acid 50 µg/ml in methanol
Catalog # MSPPMPFDA
CAS Number:  
Organic reference standard, Perfluorodecanoic acid 50 µg/ml in methanol
Catalog # MSPPMPFDA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    335-76-2
  • Description:
    Perfluorodecanoic acid 50 µg/ml in methanol
  • Formula:
    C₁₀HF₁₉O₂
  • Description:
    Perfluorodecanoic acid

Specifications

About this item