(S)-(-)-3-Hydroxy-3-phenylpropionic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36567-72-3
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00145262
:166.18 g/mol
Melting Pt:117…119 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together