Order Entry
Belgium
ContactUsLinkComponent
 
(S)-(-)-3-Hydroxy-3-phenylpropionic acid 99%
  ACRO412120010
 :  
undefined
(S)-(-)-3-Hydroxy-3-phenylpropionic acid 99%
  ACRO412120010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to off-white Crystals or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    116 to 119 °C
  • Specific optical rotation:
    -21.5° to -23.5° (20 °C, 589 nm) (c=4, CH3OH)
  • Titration with NaOH:
    ≥98.5 %

 

 

 :
 :166.18 g/mol
 :116…119 °C
 :Refrigerator
 :36567-72-3
 :C₉H₁₀O₃
 :MFCD00145262