(S)-(+)-2-Hydroxy-2-phenylpropionic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13113-71-8
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00067700
:166.18 g/mol
Melting Pt:115…117 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together