(R)-(+)-2-Hydroxy-3-phenylpropionic acid
Catalog # MOLE49727033-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:166.1739 g/mol
EINECS:230-803-3
CAS Number:7326-19-4
Formula:C₉H₁₀O₃
MDL Number:MFCD00078062