(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7326-19-4
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00078062
:166.1739 g/mol
Melting Pt:121…125 °C
Storage Temperature:Ambient
EINECS:230-803-3
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:121 to 125 °C
- Specific optical rotation:+17° to +20° (20 °C, 589 nm) (c=1, CH3OH)
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together