Calmagite
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3147-14-6
:
Formula:C₁₇H₁₄N₂O₅S
MDL Number:MFCD00011656
:358.37 g/mol
Merck Index:14,1718
Melting Pt:>330 °C
EINECS:221-563-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together