
1-(4-Nitrophenylazo)-2-naphthol, Sigma-Aldrich®
Catalog # 100994-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance (colour):Red
- Appearance (form):Powder
- Infrared spectrum:Conforms to structure
- Wavelength in 0.008 g/L, Toluene:485 - 491 nm
- Extinction coefficient:≥21.000
- Wavelength:325 - 331 nm
- Carbon:61.6 - 69.5%
- Nitrogen:13.4 - 15.2%
- TLC consistent with control:Conforms
- Solubility (turbidity) c = 1 mg/ml, acetone:Clear to slightly hazy
- Solubility (colour) orange to red :Conforms to requirements
Specification Test Results
About this item
:
:293.28 g/mol
Melting Pt:248…252 °C (lit.)
:M-Clarity™ quality level = MQ100
EINECS:229-093-8
:GHS07
CAS Number:6410-10-2
Formula:C₁₆H₁₁N₃O₃
MDL Number:MFCD00003908