Order Entry
Belgium
ContactUsLinkComponent
2-Sulphobenzoic acid monoammonium salt for synthesis, Sigma-Aldrich®
Irritant
2-Sulphobenzoic acid monoammonium salt for synthesis, Sigma-Aldrich®
Catalog # 8.21600.0025
Supplier:  Merck
CAS Number:  
Irritant
2-Sulphobenzoic acid monoammonium salt for synthesis, Sigma-Aldrich®
Catalog # 8.21600.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    274 - 277 °C
  • Molar Mass:
    219.22 g/mol

Specification Test Results

About this item

CAS Number:6939-89-5
 :
Formula:C₇H₉NO₅S
MDL Number:MFCD00040590
 :M-Clarity™ quality level = MQ200
EINECS:230-077-8
 :GHS07
Storage Temperature:Ambient