1-Methylimidazole-4-sulphonyl chloride
Catalog # MAYBKM06428.0500
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:180.61 g/mol
CAS Number:137049-00-4
Formula:C₄H₅ClN₂O₂S
MDL Number:MFCD00068060