2-Picoline-[D₇] (98% D)
Supplier: LGC Standards PROMOCHEM
Jump to:
About this item
Synonyms: 2-Picoline-[²H₇]
- CAS Number:93951-93-0
- Formula:C₆D₇N
2 Options Available Below
Product Details & Documents
Synonyms: 2-Picoline-[²H₇]
- CAS Number:93951-93-0
- Formula:C₆D₇N



