Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, HyperSep™ Diol
Solid phase extraction cartridges, HyperSep™ Diol
   Sustainable Options Available
Solid phase extraction cartridges, HyperSep™ Diol
   Sustainable Options Available
Certificates
Solid phase extraction cartridges, HyperSep™ Diol
Solid phase extraction cartridges, HyperSep™ Diol
9 Options Available Below
Product Details & Documents
A short alkyl chains with polar functional groups are ideal for the isolation of polar compounds.
  • Normal phase extraction of polar compounds
  • Purification of polar compounds
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Product Family Options
Product Information
  • Column Format
    Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • Cartridge
    Diol
    40 - 60 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 322,00 €
    322,00 €
    1 * 100 ST
       322,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HYPE60108-571
  • Cartridge
    Diol
    40 - 60 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 328,00 €
    328,00 €
    1 * 100 ST
       328,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HYPE60108-572
  • Cartridge
    Diol
    40 - 60 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 260,00 €
    260,00 €
    1 * 50 ST
       260,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HYPE60108-573
  • Cartridge
    Diol
    40 - 60 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 262,00 €
    262,00 €
    1 * 50 ST
       262,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HYPE60108-574
  • Cartridge
    Diol
    40 - 60 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 260,00 €
    260,00 €
    1 * 30 ST
       260,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HYPE60108-575
  • Cartridge
    Diol
    40 - 60 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 236,00 €
    236,00 €
    1 * 30 ST
       236,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HYPE60108-576
  • Cartridge
    Diol
    40 - 60 µm
    15 ml
    2000 mg
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 275,00 €
    275,00 €
    1 * 20 ST
       275,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HYPE60108-755
  • Cartridge
    Diol
    40 - 60 µm
    25 ml
    5000 mg
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 302,00 €
    302,00 €
    1 * 20 ST
       302,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HYPE60108-756
  • Cartridge
    Diol
    40 - 60 µm
    75 ml
    10000 mg
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 335,00 €
    335,00 €
    1 * 10 ST
       335,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HYPE60108-757
96 well plates and individual wells
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 10 mg
    96-well plate, HyperSep™ Diol
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 488,00 €
    488,00 €
    1 * 1 ST
       488,00 €
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 25 mg
    96-well plate, HyperSep™ Diol
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 537,00 €
    537,00 €
    1 * 1 ST
       537,00 €
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 100 mg
    96-well plate, HyperSep™ Diol
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 569,00 €
    569,00 €
    1 * 1 ST
       569,00 €
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 50 mg
    96-well plate, HyperSep™ Diol
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 567,00 €
    567,00 €
    1 * 1 ST
       567,00 €