Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Dispenser Boxes

Dispenser Boxes

Dispenser for speedy and clean cutting of Parafilm® and Dura Seal®, made of ABS, vert, 171×120×144 mm
Catalog # ROTHHY13.1
Marketsource
Supplier:   Roth Carl
Dispenser Boxes
Catalog # ROTHHY13.1
Supplier:  Roth Carl
Marketsource
Dispenser Boxes

Dispenser for speedy and clean cutting of Parafilm® and Dura Seal®, made of ABS, vert, 171×120×144 mm

Price
71,35 €
Log in for availability
1 * 1 ST
Product Details & Documents

Dispenser, made of ABS, for PARAFILM® sealing film.

  • Store, dispense, and cut Parafilm®, tape, labels, and Tough-Tags®
  • Designed with a safety razor that slides and cuts with no danger to fingers
  • Dispenser holds one or two 50 mm (2") rolls, or one 100 mm (4") roll
Specifications
  • Couleur:
    vert
  • Désignation:
    Dispenser for speedy and clean cutting of Parafilm® and Dura Seal®, made of ABS
  • l×p×h:
    171×120×144 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.