Order Entry
Belgium
ContactUsLinkComponent
VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
Catalog # 662-1659
Supplier:  VWR Collection
undefined
VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
Catalog # 662-1659
Supplier:  VWR Collection

Specifications

  • Temperature range:
    Auto: −5,0...+105,0 °C; manual: −25...+130 °C
  • Description:
    pH-meter pH 1100 H, pHenomenal®
  • Model:
    pH 1100 H
  • Temperature accuracy:
    ±0,2 °C
  • Temperature resolution:
    0,1 °C
  • Automatic buffer recognition:
    VWR® Chemicals TITRINORM® buffers at 25 °C: 4,00/6,98/9,94
    NIST/DIN 19266 (1,68/4,00/6,86/9,18/12,54)
    Technical buffers at 25 °C: 4,00/7,00/10,00
  • Compliance:
    IP 67/GLP
  • Connections:
    BNC for pH/banana for temperature
  • Data logging capacity:
    5000 data sets
  • Data logging - log function:
    Selectable between 1 min and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
  • Display:
    Graphic LCD with LED backlight
  • Outputs:
    Waterproof USB connection (slave, bidirectional)
  • Power supply:
    4× AA batteries
  • Temperature compensation:
    Auto/Manual
  • mV accuracy:
    ±0,3/±1
  • mV range:
    ±1200,0/±2500
  • mV resolution:
    0,1/1
  • pH accuracy:
    ±0,005 pH ±1 digit
  • pH calibration:
    Up to 5 points
  • pH range:
    −2,000...+20,000
  • pH resolution:
    0,1/0,01/0,001 (selectable)
  • Pk:
    1 SET

Specifications

About this item

This user friendly, portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements in field applications. Complete with waterproof USB output for connection to a computer. GLP compliant.

  • Large graphic LCD with continuous LED backlight shows both pH/mV and temperature value simultaneously
  • Data logging function selectable between 1 min and 1 h
  • 5000 data set memory capacity with automatic storing; additional 200-point memory for manual storage
  • Automatic power shut-off: Adjustable from 10 min to 24 h
  • Automatic buffer recognition of DIN and NIST buffers (1,68 / 4,00 / 6,86 / 9,18 / 12,54)
  • Three additional technical buffers at 25 °C (4,00 / 7,00 / 10,00) in memory (pH 10,00 not included in delivery)
  • Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
  • Low energy consumption: 4× AA batteries for over 1000 operating hours
Ordering information: Ready to use pH portable set pH 1100 H includes instrument in robust plastic case, gel epoxy pH electrode with built-in temperature sensor 111 (1 m cable), starter kit (two buffers (4,00 + 7,00), one plastic beaker), stand and instruction manual (22 languages) on CD ROM.
Delivery: Ready to use portable pH kit pH 1100 H (662-1659) includes instrument in robust plastic carrying case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, gel epoxy pH electrode with built-in temperature sensor 111 (1 m cable).

Electrode

Product Information