Order Entry
Belgium
ContactUsLinkComponent
VWR® pHenomenal® pH 1100L/1100LB pH/mV/°C Meter, Bench
VWR® pHenomenal® pH 1100L/1100LB pH/mV/°C Meter, Bench
Supplier:  VWR Collection
Certificates
undefined
VWR® pHenomenal® pH 1100L/1100LB pH/mV/°C Meter, Bench
Supplier:  VWR Collection

Specifications

  • Catalog No:
  • 662-1657
  • 662-1861
  • Model:
  • pH 1100L
  • pH 1100LB
  • pH range:
  • −2,000...+20,000
  • −2,000...+20,000
  • pH resolution:
  • 0,1 / 0,01 / 0,001 (selectable)
  • 0,1 / 0,01 / 0,001 (selectable)
  • pH accuracy:
  • ±0,1 / ±0,01 / ±0,005
  • ±0,1 / ±0,01 / ±0,005
  • pH calibration:
  • Up to 5 points
  • Up to 5 points
  • Automatic buffer recognition:
  • VWR® Chemicals TITRINORM® buffers at 25 °C: 4,00 / 6,98 / 9,94
    NIST / DIN 19266 (1,68 / 4,00 / 6,86 / 9,18 / 12,54)
    Technical buffers at 25 °C: 4,00 / 7,00 / 10,00
  • VWR® Chemicals TITRINORM® buffers at 25 °C: 4,00 / 6,98 / 9,94
    NIST / DIN 19266 (1,68 / 4,00 / 6,86 / 9,18 / 12,54)
    Technical buffers at 25 °C: 4,00 / 7,00 / 10,00
  • mV range:
  • ±1200,0 / ±2500
  • ±1200,0 / ±2500
  • mV resolution:
  • 0,1 / 1 (selectable)
  • 0,1 / 1 (selectable)
  • mV accuracy:
  • ±0,3 / ±1
  • ±0,3 / ±1
  • Temperature range:
  • Auto: −5,0...+105,0 °C; manual: −25...+130 °C
  • Auto: −5,0...+105,0 °C; manual: −25...+130 °C
  • Temperature resolution:
  • 0,1/1 °C
  • 0,1/1 °C
  • Temperature accuracy:
  • ±0,2 °C
  • ±0,2 °C
  • Temperature compensation:
  • Auto / manual
  • Auto / manual
  • Display:
  • Graphic LCD with LED backlight
  • Graphic LCD with LED backlight
  • Power supply:
  • Mains or 4× AA batteries
  • Mains or 4× AA batteries
  • Compliance:
  • IP 43 / GLP
  • IP 43 / GLP
  • Data logging - log function:
  • Selectable between 1 min and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
  • Selectable between 1 min and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
  • Data logging capacity:
  • 5000 data sets
  • 5000 data sets
  • Connections:
  • BNC for pH / banana for temperature
  • BNC for pH / banana for temperature
  • Outputs:
  • USB connection (slave, bidirectional)
  • USB connection (slave, bidirectional)

Specifications

Electrodes

Product Information

Frequently Bought Together