Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lids
Embedding Cassette Lids
Lids for cassettes
Lids for cassettes
Lids for cassettes
Catalog # 216-4335
Supplier:
DELTALAB
Expand image
Lids for cassettes
Catalog # 216-4335
Supplier:
DELTALAB
Specifications
Colour:
Grey
Seq:
06
Pk:
10
Specifications
About this item
Product Details
Stainless steel lids.
Fit all standard cassettes
Product Details