Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Tissue processing cassettes, polyacetylene

Tissue processing cassettes, polyacetylene
Supplier:  DELTALAB
Certificates
Tissue processing cassettes, polyacetylene

5 Options Available Below

Product Details & Documents

These cassettes are made of polyacetyle.

  • With 34 round holes of 2 mm
  • External dimensions: 40×28×6 mm
  • Can be written on

They are resistant to the solvents used in histological processes.

 

Product Family Options

Product Information

  • Colour
    Availability
    Price
  • Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 99,50 €
    99,50 €
    1 * 1.000 ST
       99,50 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 101,00 €
    101,00 €
    1 * 1.000 ST
       101,00 €

  • Red
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST

  • White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 58,90 €
    58,90 €
    1 * 1.000 ST
       58,90 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 101,00 €
    101,00 €
    1 * 1.000 ST
       101,00 €

Lids

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Grey
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 109,00 €
    109,00 €
    1 * 10 ST
       109,00 €