Orders LinkContactUsLinkComponent
Containers, square, with screw cap
Containers, square, with screw cap
Supplier:  DELTALAB
Certificates
Containers, square, with screw cap
7 Options Available Below
Product Details & Documents
Containers and insert plugs made of HDPE, caps are made of PP.
  • Good chemical and temperature resistance
  • Tagging points on neck and cap for a tamper proof closure or for labelling
  • All models have moulded-in graduations
Translucent square containers with star-shaped caps.
Product Family Options
Product Information
  • Capacity
    W×D×H
    Colour
    Neck-Ø
    Availability
    Price
  • 25 ml
    32×34×52 mm
    Translucent white
    18 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 564,00 €
    564,00 €
    1 * 500 ST
       564,00 €
  • 50 ml
    38×38×70 mm
    Translucent white
    23 mm
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 529,00 €
    529,00 €
    1 * 400 ST
       529,00 €
  • 100 ml
    42×48×90 mm
    Translucent white
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,40 €
    1,40 €
    1 * 1 ST
       1,40 €
  • 250 ml
    57×60×110 mm
    Translucent white
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,90 €
    1,90 €
    1 * 1 ST
       1,90 €
  • 500 ml
    70×80×138 mm
    Translucent white
    44 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,50 €
    2,50 €
    1 * 1 ST
       2,50 €
  • 1000 ml
    80×103×176 mm
    Translucent white
    58 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,00 €
    4,00 €
    1 * 1 ST
       4,00 €
  • 2000 ml
    100×134×210 mm
    Translucent white
    58 mm
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 292,00 €
    292,00 €
    1 * 40 ST
       292,00 €