Microplates, 96-well, non binding
Supplier: Greiner Bio-One
Microplates, 96-well, non binding
Supplier: Greiner Bio-One
Certificates
About this item
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
7 Options Available Below
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
Specifications for Product Family
Specifications
- Catalog No:
- 738-0165
- 738-0166
- 738-0170
- 738-0172
- 738-0169
- 738-0171
- 738-0173
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
Product Information
- ColourAvailabilityPrice
- 738-016596-well microplate, U-bottom, non binding, solid bottom96-well microplate, U-bottom, non binding, solid bottomClearLog in for availability1 * 40 ST298,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0165Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-016696-well microplate, V-bottom, non binding, solid bottom96-well microplate, V-bottom, non binding, solid bottomClearLog in for availability1 * 40 ST298,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0166Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-017096-well microplate, F-bottom, non binding, chimney well, solid bottom96-well microplate, F-bottom, non binding, chimney well, solid bottomClearLog in for availability1 * 40 ST265,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0170Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-017296-well microplate, F-bottom, non binding, chimney well, solid bottom96-well microplate, F-bottom, non binding, chimney well, solid bottomWhiteLog in for availability1 * 40 ST291,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0172Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-016996-well microplate, F-bottom, non binding, chimney well, solid bottom96-well microplate, F-bottom, non binding, chimney well, solid bottomBlackLog in for availability1 * 40 ST316,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0169Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-017196-well microplate, F-bottom, non binding, chimney well, μClear® bottom96-well microplate, F-bottom, non binding, chimney well, μClear® bottomWhite/clearLog in for availability1 * 40 ST462,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0171Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 738-017396-well microplate, F-bottom, non binding, chimney well, μClear® bottom96-well microplate, F-bottom, non binding, chimney well, μClear® bottomBlack/clearLog in for availability1 * 40 ST462,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.738-0173Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact