Orders LinkContactUsLinkComponent
Wire baskets, rectangular, with handle
Wire baskets, rectangular, with handle
Wire baskets, rectangular, with handle
Supplier:  DELTALAB
Certificates
Wire baskets, rectangular, with handle
2 Options Available Below
Product Details & Documents
Made of stainless steel.
  • Features a strong woven mesh base
  • With folding handles
  • Suitable for bottles and containers
Documents
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 216-7513
    Without dividers
    Without dividers
    480×300×150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 266,00 €
    266,00 €
    1 * 1 ST
       266,00 €
  • 216-7515
    With dividers, 4 compartments
    With dividers, 4 compartments
    480×300×150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST