Orders LinkContactUsLinkComponent
Microtubes racks
Microtubes racks
Microtubes racks
Supplier:  DELTALAB
Certificates
Microtubes racks
5 Options Available Below
Product Details & Documents
Space saving racks, made of PP, designed to accommodate microtubes for freezer storage.
  • Alpha-numeric matrix allows accurate sample identification
  • Lateral handles allow easy handling of the racks
  • Use with 1,5/2,0 ml microtubes and 2 ml cryotubes
  • Suitable for freezer use down to −80 °C
  • Autoclavable
Documents
Product Family Options
Product Information
  • Colour
    No. of Holes
    For tube Ø
    W×D×H
    Availability
    Price
  • Blue
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 56,10 €
    56,10 €
    1 * 5 ST
       56,10 €
  • Green
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 56,10 €
    56,10 €
    1 * 5 ST
       56,10 €
  • Natural
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 52,90 €
    52,90 €
    1 * 5 ST
       52,90 €
  • Red
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 56,10 €
    56,10 €
    1 * 5 ST
       56,10 €
  • Yellow
    80 (5×16)
    11 mm
    225×65×27 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 56,10 €
    56,10 €
    1 * 5 ST
       56,10 €