ContactUsLinkComponent
Surgical scissors, single use, Aesculap® SUSI®
Surgical scissors, single use, Aesculap® SUSI®
Catalog # AESCBC324SU
Supplier:  Aesculap
Surgical scissors, single use, Aesculap® SUSI®
Catalog # AESCBC324SU
Supplier:  Aesculap
Specifications
  • Type:
    Straight, sharp/blunt
  • Length:
    145 mm
  • Pk:
    20
Specifications
About this item
These disposable scissors are made from a high performance polymer. The special composition of this high-grade polymer ensures an outstanding quality and provides optimal mechanical properties.
  • Single use reduces risk of cross-contamination
  • Reduction of processing costs in hospitals
  • Sterilised in a validated process
The material ensures a safe, easy and environmentally friendly removal through incineration in the regular hospital waste removal system.
Packaging: The scissors are sterile packed in single peel packs. The single peel packs are presented in dispenser boxes containing an indicated quantity of units as described in this leaflet.