ContactUsLinkComponent
Beakers, low form, borosilicate glass
  SMIT155/600
 : 155/600
Beakers, low form, borosilicate glass
  SMIT155/600
 : 155/600
Select an Option
90 mm
Select an Option
600 ml
Select an Option
125 mm
Select an Option
09
Select an Option
1
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Autoclavable
Ideal for general laboratory use. With spout and white graduations.
Frequently Bought Together