ContactUsLinkComponent
Repeating adapter for syringes
Repeating adapter for syringes
Catalog # SGEA031930
Repeating adapter for syringes
Catalog # SGEA031930
Specifications
  • Description:
    RAX repeating adapter, for syringes from 500 nl - 500 μl
  • Pk:
    1
Specifications
About this item
The RAX repeating adapter improves precision and reproducibility when repeatedly dispensing the same volume manually. It is suitable for syringes (except those with plunger stops, or diluter/dispenser syringes) and can be used on syringe barrels (Ext.Ø 6,5 and 8 mm).
  • Easy to use finger grips and can be set to any desired volume
  • Used as added protection against both plunger blow out at elevated pressures and plunger bending
The flat sides allow it to sit securely on the bench with the needle in a raised position helping to prevent damage from rolling. The double-ended tool facilitates assembly, it can easily and quickly be installed and if required it can readily be removed, replaced or swapped between syringes.
Delivery: Supplied with double-ended tool.