Order Entry
Belgium
Orders LinkContactUsLinkComponent
Gas cylinder storage boxes

Gas cylinder storage boxes

Gas cylinder storage box, with intermediate shelves and standard cylinder lock, for 10×11 kg cylinders, 1-door, perforated, 840×690×1475 mm
Catalog # DUEH7.90.100
Supplier:   DUPERTHAL
Gas cylinder storage boxes
Catalog # DUEH7.90.100
Supplier:  DUPERTHAL
Gas cylinder storage boxes
Gas cylinder storage boxes

Gas cylinder storage box, with intermediate shelves and standard cylinder lock, for 10×11 kg cylinders, 1-door, perforated, 840×690×1475 mm

Price
715,71 €
Log in for availability
1 * 1 ST
Product Details & Documents

Storage box is used for storing gas cylinders outdoors in accordance with the regulations.

  • Galvanised sheet steel
  • Cylinder lock as standard fitting
  • Disassembled upon delivery – easy to assemble

Certifications: Conforms with TRGS 510.

Delivery information: Supplied with intermediate shelf.

Specifications
  • Description:
    Gas cylinder storage box, with intermediate shelves and standard cylinder lock, for 10×11 kg cylinders
  • W×D×H:
    840×690×1475 mm
  • Doors:
    1-door, perforated
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.