Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bromophenol blue ACS

Bromophenol blue ACS
Supplier:  VWR Chemicals
Certificates
Bromophenol blue ACS

2 Options Available Below

Product Details & Documents

A tracking dye for nucleic acid and protein gels. Also a pH indicator (yellow to blue, range 3,0 to 4,6).

Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:13,01429
  • Melting Pt:273 °C
  • Density:1 g/cm³ (20 °C)
  • EINECS:204-086-2
Specifications for Product Family

Specification Test Results

  • Clarity of Solution:
    Pass
  • Visual Transition Interval (pH 3.0-4.6):
    Pass
Product Family Options

Product Information