HomeTubingChromatography TubingFused silica tubing, Upchurch Scientific®
Made from high quality, polyimide-clad fused silica.
This tubing is ideal for micro-scale analyses such as micro and nano- HPLC and capillary electrophoresis because of the tight tolerances of fused silica’s inner diameters.
: EUTECH
: LEICA MICROSYSTEMS
: EUROMEX
: VWR Collection
: UPCHURCH SCIENTIFIC
: VISION ENGINEERING
: OLYMPUS OPTICAL