Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, HyperSep™ C8
Solid phase extraction cartridges, HyperSep™ C8
Solid phase extraction cartridges, HyperSep™ C8
Certificates
Solid phase extraction cartridges, HyperSep™ C8
Solid phase extraction cartridges, HyperSep™ C8
8 Options Available Below
Product Details & Documents
HyperSep™ C8 cartridges, less retentive alternative to C18 for polar and nonpolar compounds.
  • Well suited for method requiring less retention than C18
  • For moderately polar analytes
  • Potential for polar interactions greater than with C18
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Product Family Options
Product Information
  • Column Format
    Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • Cartridge
    C8
    40 - 60 µm
    1 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 190,00 €
    190,00 €
    1 * 50 ST
       190,00 €
  • Cartridge
    C8
    40 - 60 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 244,00 €
    244,00 €
    1 * 100 ST
       244,00 €
  • Cartridge
    C8
    40 - 60 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 262,00 €
    262,00 €
    1 * 100 ST
       262,00 €
  • Cartridge
    C8
    40 - 60 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 177,00 €
    177,00 €
    1 * 50 ST
       177,00 €
  • Cartridge
    C8
    40 - 60 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 169,00 €
    169,00 €
    1 * 30 ST
       169,00 €
  • Cartridge
    C8
    40 - 60 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 197,00 €
    197,00 €
    1 * 30 ST
       197,00 €
  • Cartridge
    C8
    40 - 60 µm
    15 ml
    2000 mg
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 226,00 €
    226,00 €
    1 * 20 ST
       226,00 €
  • Cartridge
    C8
    40 - 60 µm
    75 ml
    10000 mg
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 293,00 €
    293,00 €
    1 * 10 ST
       293,00 €
96 well plates and individual wells
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 50 mg
    96-well plate, HyperSep™ C8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 524,00 €
    524,00 €
    1 * 1 ST
       524,00 €
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 100 mg
    96-well plate, HyperSep™ C8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 537,00 €
    537,00 €
    1 * 1 ST
       537,00 €
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 10 mg
    96-well plate, HyperSep™ C8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 494,00 €
    494,00 €
    1 * 1 ST
       494,00 €
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 25 mg
    96-well plate, HyperSep™ C8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 517,00 €
    517,00 €
    1 * 1 ST
       517,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.