Order Entry
Belgium
ContactUsLinkComponent
VWR® Q Path® DC3, Decalcifier Rapid-Acting
VWR® Q Path® DC3, Decalcifier Rapid-Acting
Catalog # 09128300.
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Q Path® DC3, Decalcifier Rapid-Acting
Catalog # 09128300.
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Description:
    Decalcifier DC3 rapid

Specifications

About this item

Contains a mixture of hydrochloric acid and alcohols with some EDTA.

  • Decalcify larger tissues (already fixated)
  • Recommended volume is 20× the fragment volume

Ready to use for applications in histology and pathology laboratories.