Order Entry
Belgium
ContactUsLinkComponent
Bottle top vacuum filtration systems, AcroVac™
Bottle top vacuum filtration systems, AcroVac™
Catalog # PALLAVFP02S
Bottle top vacuum filtration systems, AcroVac™
Catalog # PALLAVFP02S

Specifications

  • Capacity:
    250 ml
  • Description:
    Filter unit with Supor® membrane, 0,2 μm, sterile, 250 ml
  • Seq:
    01
  • Membrane Ø:
    102 mm
  • Pk:
    12

Specifications

About this item

These filter units are designed for fast vacuum filtration. The low centre of gravity improves the filtration system stability and reduces the risk of system tipping during the filtration process.

  • Curved sides moulded into the reservoir
  • Finger tabs on the funnel base and storage cap allow for easy removal and tightening

PS funnel; copolyester collection reservoir; PP lid, funnel base, storage cap and vacuum port; Supor® PES membrane.

Applications: Cell culture media filtration, buffer filtration, tissue culture additive filtration, clarification of biologicals.

Systems are non-cytotoxic and meet the requirements of the Elution Test, ISO 10993-5, 1999.

Recommended/max. operating vacuum: 380/510 mm Hg
Max. temperature: 37 °C

Samples of 0,2 μm pore size systems retain a minimum of 107 cfu/cm² of Brevundimonas diminuta per modified ASTM F838; current revision.

Packaging: Individually bagged and sterilised by gamma irradiation.