Order Entry
Belgium
Orders LinkContactUsLinkComponent
Lateral Flow test for Campylobacter, Singlepath® Campylobacter

Lateral Flow test for Campylobacter, Singlepath® Campylobacter

Singlepath® Campylobacter, 25 Tests
Catalog # 1.04143.0001
Supplier:   Merck
Lateral Flow test for Campylobacter, Singlepath® Campylobacter
Catalog # 1.04143.0001
Supplier:  Merck
CAS Number:  
Lateral Flow test for Campylobacter, Singlepath® Campylobacter
Lateral Flow test for Campylobacter, Singlepath® Campylobacter

Singlepath® Campylobacter, 25 Tests

CAS Number:  
Price
252,00 €
Log in for availability
1 * 25 Tests
Product Details & Documents

Campylobacteriosis is an infection caused by Campylobacter, most commonly C. jejuni. It produces an inflammatory, sometimes bloody, diarrhea or dysentery, including cramps, fever and abdominal pain. The debilitating neurological disorder, Guillain-Barre Syndrome (GBS), as well as reactive arthritis have also been associated with recent infections with certain C. jejuni strains. C. lari and the emerging pathogen, C. upsaliensis, have also been reported in a small percentage of cases of human Campylobacter infection. Campylobacter spp. are highly infectious - as few as 500 bacteria can cause illness. There is an increasing demand for Campylobacter testing in food to meet regulations regarding Campylobacter levels in poultry. The majority of Campylobacter spp. are relatively metabolically inactive, making identification based on biochemical characteristics difficult. Currently, the most commonly used techniques to test food products for Campylobacter are traditional methods based on culture media. Innovative, rapid tests make screening for pathogenic organisms easier. The one-step format reduces the possibility of errors during handling.

  • Easy to use: simply add sample and read off the result
  • Safe: clear and distinctive positive or negative test results with built-in positive control
  • Reliable: as sensitive as the official culture media method
  • Economical: rapid results save labour and inventory costs

Certifications: According AOAC-RI

Documents

Specifications

Specifications

  • Pk:
    25 Tests
  • Description:
    Singlepath® Campylobacter

Specifications

Accessories

Product Information

  • Catalog #
    Standards
    Pk
    Availability
    Price
  • GranuCult™ Bolton broth (base)
    GranuCult™ Bolton broth (base)
    1.00068.0500
    ISO 10272
    500 g
    Log in for availability
    1 * 500 g - 280,00 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Bolton broth selective supplement
    Bolton broth selective supplement
    1.00079.0010
    10 Vial
    Log in for availability
    1 * 10 VIA - 417,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.