



n-Heptane
Supplier: Merck
Certificates
About this item
CAS Number:142-82-5
:
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
:100.20 g/mol
Merck Index:13,04679
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–90.6 °C
Density:0.6838 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:–4 °C
:GHS02
:GHS08
:GHS07
:GHS09
Frequently Bought Together