Single-sided step ladders
About this item
Ascendable on one side, grooved steps with a depth of 80 mm with durable 'step to side' rail connection, non slip nivello® ladder feet. High standing safety ensured by conical construction.
- Connection on both sides by two detachable metal struts
- Spacing between steps: 235 mm
- Ladder inclination: 20°
- Non slip nivello® ladder shoes
- Ergopad grip zone
- Safe cap joint
- Durable 'step to side' rail connection
Certifications: GS tested; in accordance with European standard DIN EN 14183.
Action manual BGI 694 and the applicable accident prevention regulations.
4 Options Available Below
Product Details & Documents
Ascendable on one side, grooved steps with a depth of 80 mm with durable 'step to side' rail connection, non slip nivello® ladder feet. High standing safety ensured by conical construction.
- Connection on both sides by two detachable metal struts
- Spacing between steps: 235 mm
- Ladder inclination: 20°
- Non slip nivello® ladder shoes
- Ergopad grip zone
- Safe cap joint
- Durable 'step to side' rail connection
Certifications: GS tested; in accordance with European standard DIN EN 14183.
Action manual BGI 694 and the applicable accident prevention regulations.
Documents
VWR® For Your Space
Product Literature
Product Family Options
Product Information
- No. of stepsWorking reach up to approx.HeightPlatform heightOutside widthW×D×HWeightAvailabilityPrice
- 139-0769Step ladder with 3 steps
- 139-0650Step ladder with 3 steps and four self-locking fixed castorsStep ladder with 3 steps and four self-locking fixed castors32750 mm1370 mm700 mm470 mm8,1 kgLog in for availability1 * 1 ST
- GUEN41503Step ladder with 3 steps and slip resistant clip-step insertsStep ladder with 3 steps and slip resistant clip-step inserts33000 mm670 mm500×700×1600 mm7,3 kgLog in for availability1 * 1 ST
- GUEN41504Step ladder with 4 steps and slip resistant clip-step insertsStep ladder with 4 steps and slip resistant clip-step inserts43250 mm1830 mm1170 mm530 mm8,5 kgLog in for availability1 * 1 ST