Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Formaldehyde 30% in aqueous solution, Q Path®

Plastic container, 5 L
Catalog # 11699031.
Toxic
Corrosive
Health
Supplier:   VWR Chemicals
Formaldehyde 30% in aqueous solution, Q Path®
Catalog # 11699031.
Supplier:  VWR Chemicals
CAS Number:  50-00-0
Formaldehyde 30% in aqueous solution, Q Path®
Formaldehyde 30% in aqueous solution, Q Path®

Plastic container, 5 L

CAS Number:  50-00-0
Price
30,50 €
Log in for availability
1 * 5 L
Product Details & Documents

Formalin 30% fixative for tissue samples.

 : Store this product at ambient temperature (above 15 °C), because it may polymerise at temperatures below 9 °C. Presence of polymers does not affect product properties. Precipitate can be removed by filtration.

Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
Specifications

Specification Test Results

  • Assay:
    28.5 - 31.5 %
  • Appearance:
    Clear colourless liquid
  • Density (20/4):
    1.055 - 1.085
  • Ashes:
    Max. 0.08 %
  • Heavy metals:
    Max. 8 ppm
  • Formic acid:
    Max. 0.08 %
  • Methanol:
    4.0 - 12.5 %
  • Fe (Iron):
    Max. 8 ppm

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L

Specifications