Order Entry
Belgium
ContactUsLinkComponent
Replacement needles for LC XCHANGE® syringes
Replacement needles for LC XCHANGE® syringes
Catalog # 548-1804
Replacement needles for LC XCHANGE® syringes
Catalog # 548-1804

Specifications

  • Ext.Ø:
    0,71 mm
  • Gauge:
    22
  • Type of tip:
    LC
  • Description:
    Replacement needles for XCHANGE® LC syringes, 1000 µl
  • Length:
    56 mm
  • Seq:
    37
  • Pk:
    5

Specifications

About this item