ContactUsLinkComponent
Mounting plate for pipette canisters
  BELC1288-11111
undefined
Mounting plate for pipette canisters
  BELC1288-11111
  • Description:
    Mounting plate, 90×90 mm, for use with 38 mm OD glass pipette canisters
  • Pk:
    6
Mounting plate for pipette canisters, made from stainless steel.
  • Stabilises pipette canisters during use
  • Can be used with 38 mm Ext.Ø glass pipette canisters