Order Entry
Belgium
ContactUsLinkComponent
Haematoxylin solution (Harris), Q Path® for microscopy
Irritant
Haematoxylin solution (Harris), Q Path® for microscopy
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Haematoxylin solution (Harris), Q Path® for microscopy
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Colour of solution:
    Purplish red
  • Density (20/4):
    1,053 - 1,059
  • pH (20°C):
    2,2 - 2,6
  • Absorbance (0.5 %):
    553 - 560 nm

Specification Test Results

Frequently Bought Together