Order Entry
Belgium
ContactUsLinkComponent
 
 
Haematoxylin solution (Mayer), Q Path® for microscopy
 
undefined
 
Haematoxylin solution (Mayer), Q Path® for microscopy
 

 

  • Appearance:
    Clear liquid
  • Colour of solution:
    Purplish red
  • Density (20/4):
    1,05 - 1,07
  • pH (20°C):
    2,2 - 2,6
  • Absorbance (0.1 %):
    559 - 561 nm

 

Frequently Bought Together