Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Formalin 10%, Q Path®, buffered
 
undefined
 
 
Formalin 10%, Q Path®, buffered
 

 

  • Appearance:
    Colourless liquid
  • Test formaldehyde:
    Passes test
  • Density (20/4):
    1,012 - 1,014
  • pH (20°C):
    6,8 - 7,2

 

Frequently Bought Together