ContactUsLinkComponent
VWR® Lysis Buffer, Hypotonic
VWR® Lysis Buffer, Hypotonic
Catalog # M334-100ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Lysis Buffer, Hypotonic
Catalog # M334-100ML
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Hyptonic lysis buffer
Specifications
About this item
Detergent-free buffer for isolation of proteins from tissue culture cells. Cell lysis and the subsequent isolation of the crude cytoplasmic fraction completed in less than 30 m.
The protocol is compatible with a variety of downstream applications including Western Blotting and ELISA procedures, as well as protein assays, which are sensitive to the presence of detergents in the lysis buffer.