ContactUsLinkComponent
VWR® Buffer, MOPS-SDS, 20X, UltraPure Grade
VWR® Buffer, MOPS-SDS, 20X, UltraPure Grade
Catalog # K855-500ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Buffer, MOPS-SDS, 20X, UltraPure Grade
Catalog # K855-500ML
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    MOPS-SDS buffer, 20X concentrate
Specifications
About this item
Useful for high resolution of proteins on neutral pH SDS-PAGE gels.
  • Ultra-pure grade
  • Convenient concentrations and packaging
  • Good quality, consistency and resolution