Order Entry
Belgium
ContactUsLinkComponent
VWR® Phosphate Buffered Saline (PBS) with 0,05% Tween®20
VWR® Phosphate Buffered Saline (PBS) with 0,05% Tween®20
Catalog # E715-1L
Fournisseur:  VWR Chemicals
Numéro CAS:  
undefined
VWR® Phosphate Buffered Saline (PBS) with 0,05% Tween®20
Catalog # E715-1L
Fournisseur:  VWR Chemicals
Numéro CAS:  

Spécifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    PBS-TWEEN pH 7.5, ultrapure
  • Seq:
    02

Spécifications

À propos de cet article

Phosphate Buffered Saline (PBS) with 0,05% Tween® 20, pH 7,5 is a ready-to-use 1× concentration.