Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Gram's decolourising solution

1 L
Catalog # 911530ZA
Supplier:   VWR Chemicals
Gram's decolourising solution
Catalog # 911530ZA
Supplier:  VWR Chemicals
CAS Number:  
Gram's decolourising solution
Gram's decolourising solution

1 L

CAS Number:  
Price
38,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Gram's Decoloriser Solution
  • MDL Number:MFCD00240361
Specifications

Specifications

  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.