Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lugol's iodine solution
Lugol's iodine solution stabilised
Lugol's iodine solution stabilised
Lugol's iodine solution stabilised
Catalog # 911520ZA
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Lugol's iodine solution stabilised
Catalog # 911520ZA
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation