Orders LinkContactUsLinkComponent
VWR® Q Path® Formaldehyde Neutraliser
Formaldehyde neutralizer Q Path®, Plastic bucket
  00699030.
VWR® Q Path® Formaldehyde Neutraliser
  00699030.
VWR® Q Path® Formaldehyde Neutraliser
VWR® Q Path® Formaldehyde Neutraliser
Formaldehyde neutralizer Q Path®, Plastic bucket
128,00 €
1 * 5 kg
Acidic neutraliser and pre-cleaner based on organic acid.
125 g of neutraliser are used to neutralise one L of formalin 4%. Agitate, wait 24 H and the process is completed.
  • Description:
    Formaldehyde neutralizer Q Path®
  • Pack type:
    Plastic bucket