
Eosin Y (yellowish) alcoholic solution, Q Path® for microscopy
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:17372-87-1
:
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040
ADR:3,III
:691.86 g/mol
EINECS:241-409-6
UN:1170
:GHS02
Storage Temperature:Ambient
:1
Specification Test Results
- Appearance:Clear liquid
- Colour of solution:Orange
- Density (20/4):0.920 - 0.940
- pH (20°C):4.9 - 5.7
- Absorbance (0.1 %):514 - 518 nm
Specification Test Results
Related Information
Frequently Bought Together