Copper (II) ethylenediamine 1 mol/l, VOLUSOL®
Catalog # 5761.5000
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Density (20/4):1.096 - 1.102
- Copper:0.98 - 1.02 mol/l
- Mol proportion (Ethylenediamine/Cu):1.90 - 2.10
Specification Test Results
About this item
:
:217.76 g/mol
EINECS:238-597-7
UN:1761
ADR:8 (6.1),II
:GHS05
CAS Number:14552-35-3
Formula:Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MDL Number:MFCD00274628