

Potassium dichromate 6.8253 g/L in aqueous solution
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7778-50-9
:
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367
:294.18 g/mol
Storage Temperature:Ambient
EINECS:231-906-6
UN:3287
ADR:6.1,III
:GHS06
:GHS08
Frequently Bought Together