Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methyl red in aqueous solution

Methyl red in aqueous solution

Supplier:   Lovibond
Methyl red in aqueous solution
Supplier:  Lovibond
Certificates
Methyl red in aqueous solution

2 Options Available Below

Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • EINECS:207-776-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 19,80 €
    19,80 €
    1 * 25 mL
       19,80 €

  • Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 36,50 €
    36,50 €
    1 * 100 mL
       36,50 €